Learn More
Specifications
Specifications
| Gene ID (Entrez) | 54476 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-56891 |
| Gene Alias | EC 6.3.2.-, ring finger protein 216 Ubiquitin-conjugating enzyme 7-interacting protein 1, Triad domain-containing protein 3, TRIAD3Zinc finger protein inhibiting NF-kappa-B, U7I1, UBCE7IP1ubiquitin conjugating enzyme 7 interacting protein 1, ZINE3 ubiquitin-protein ligase RNF216 |
| Gene Symbol | RNF216 |
| Quantity | 0.1 mg |
| Research Category | Innate Immunity |
| Storage Buffer | Peptide dissolved in dH20. Contains no BSA. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
