Learn More
Quidel Triage MeterPro
SDP
Catalog No. 13-982-25
Click to view available options
:
1 meter
Catalog No. 13-982-25
Supplier: Quidel 55070
Only null left
Add to Cart
Triage MeterPro is a point of care instrument with a variety of immunoassays and the ability to use whole blood, plasma, or urine.
  • Barcode reader for patient and operator ID with alphanumeric capability
  • ID and QC lockouts including unauthorized users
  • Test Select™ customizable test panels: run only what you need
  • Interface with any LIS solution
  • Multiple comprehensive connectivity options
Specifications
Type Triage MeterPro
Sample Type Whole Blood, Plasma, or Urine
Analysis Time 15 to 20 min.
Measurement Method Immunoassay
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.