Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triallyl phosphite

Catalog No. AA1748418
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 102-84-1 | C9H15O3P | 202.19 g/mol

Triallyl phosphite is employed as a radical precursor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-84-1
Molecular Formula C9H15O3P
Molecular Weight (g/mol) 202.19
MDL Number MFCD00014948
InChI Key KJWHEZXBZQXVSA-UHFFFAOYSA-N
Synonym triallyl phosphite, allyl phosphite, triallylphosphite, triallyloxyphosphine, phosphorous acid, tri-2-propenyl ester, allyl phosphite, c3h5o 3p, phosphorous acid, triallyl ester, allyl phosphite 6ci,7ci, phosphorous acid, tri-2-propen-1-yl ester, tris prop-2-en-1-yl phosphite
PubChem CID 66893
IUPAC Name tris(prop-2-enyl) phosphite
SMILES C=CCOP(OCC=C)OCC=C

Specifications

Color Colorless
Boiling Point 84°C (12 mmHg)
Flash Point 86°C (187°F)
Odor Strong
Linear Formula (CH2=CHCH2O)3P
Quantity 50 g
UN Number UN3278
Beilstein 1705535
Sensitivity Air and moisture sensitive
Solubility Information Not miscible or difficult to mix.
Formula Weight 202.19
Physical Form Liquid
Chemical Name or Material Triallyl phosphite
Show More Show Less
Hazard Category H301-H312
Hazard Statement GHS H Statement
H301-H312
Toxic if swallowed.
Harmful in contact with skin.
Precautionary Statement P264b-P270-P280-P301+P310-P302+P352-P312-P330-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOPHOSPHORUS COMPOUND, LIQUID, TOXIC, N.O.S.
EINECSNumber 203-060-8
RTECSNumber TH1772500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.