Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triallylamine, 97%

Catalog No. p-7081900
Click to view available options
Quantity:
250 g

CAS: 102-70-5 | C9H15N | 137.226 g/mol

It is employed in the production of other chemicals. It is also mainly applied in organic synthesis and resin modifiers. It is used in the cross linking of high absorbent and the intermediates of ion exchange resin. It can also be employed in producing polyester activator and the evocating agent of butadiene polymerization.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the production of other chemicals. It is also mainly applied in organic synthesis and resin modifiers. It is used in the cross linking of high absorbent and the intermediates of ion exchange resin. It can also be employed in producing polyester activator and the evocating agent of butadiene polymerization.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from strong oxidizing agents and acids. Protect against electrostatic charges and keep away from ignition sources.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 102-70-5
Molecular Formula C9H15N
Molecular Weight (g/mol) 137.226
MDL Number MFCD00026093
InChI Key VPYJNCGUESNPMV-UHFFFAOYSA-N
Synonym triallylamine, tris 2-propenyl amine, 2-propen-1-amine, n,n-di-2-propenyl, triallyl amine, unii-b6n19xc04r, ccris 4876, n,n-diallylprop-2-en-1-amine, n,n-di-2-propenyl-2-propen-1-amine, ch2=chch2 3n, 4-04-00-01061 beilstein handbook reference
PubChem CID 7617
IUPAC Name N,N-bis(prop-2-enyl)prop-2-en-1-amine
SMILES C=CCN(CC=C)CC=C

Specifications

Melting Point -70°C
Density 0.79
Boiling Point 150°C to 151°C
Flash Point 31°C (87°F)
Odor Amine-like
Refractive Index 1.451
Quantity 250 g
UN Number UN2610
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 137.23
Percent Purity 98%
Chemical Name or Material Triallylamine
Show More Show Less
Hazard Category H226-H302+H332-H311-H314-H335
Hazard Statement GHS H Statement
H311-H314-H318-H226-H302-H332
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: TRIALLYLAMINE
EINECSNumber 203-048-2
RTECSNumber XX5950000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.