Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triamcinolone acetonide, Analytical Standard, ≥99%, MilliporeSigma™ Supelco™

Catalog No. 111015453
Click to view available options
Quantity:
250 mg
1 g

Tandem Mass Spectrometry data independently generated by Scripps Center for Metabolomics is available to view or download in PDF. T6501.pdf Tested metabolites are featured on Scripps Center for Metabolomics METLIN Metabolite Database. To learn more, visit sigma.com/metlin.

TRUSTED_SUSTAINABILITY

Specifications

Packaging Glass Bottle
CAS 76-25-5
Physical Form Neat
Linear Formula C24H31FO6
MDL Number MFCD00056834
Synonym 9 alpha-Fluoro-11 beta,16 alpha,17 alpha,21-tetrahydroxy-1,4-pregnadiene-3,20-dione 16,17-acetonide; 9 alpha-Fluoro-16 alpha-hydroxyprednisolone 16 alpha,17 alpha-acetonide
Quantity 1 g
Formula Weight 434.50
Percent Purity ≥99%
Grade Analytical Standard
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.