Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA562821
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA562821 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA562821 Supplier Invitrogen™ Supplier No. PA562821
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: EACTDMKREY DQCFNRWFAE KFLKGDSSGD PCTDLFKRYQ QCVQKAIKEK EIPIEGLEFM GHGKE Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.

Involved in the modulation of the mitochondrial apoptotic pathway by ensuring the accumulation of cardiolipin (CL) in mitochondrial membranes. In vitro, the TRIAP1:PRELID1 complex mediates the transfer of phosphatidic acid (PA) between liposomes and probably functions as a PA transporter across the mitochondrion intermembrane space to provide PA for CL synthesis in the inner membrane. Likewise, the TRIAP1:PRELID3A complex mediates the transfer of phosphatidic acid (PA) between liposomes (in vitro) and probably functions as a PA transporter across the mitochondrion intermembrane space (in vivo). Mediates cell survival by inhibiting activation of caspase-9 which prevents induction of apoptosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIAP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.20 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene TRIAP1
Gene Accession No. O43715
Gene Alias 15E1.1; 1810015M01Rik; AU020874; AU043831; HSPC132; MDM35; mitochondrial distribution and morphology 35 homolog; P53CSV; p53-inducible cell-survival factor; Protein 15E1.1; TP53 regulated inhibitor of apoptosis 1; TP53-regulated inhibitor of apoptosis 1; Triap1; WF-1
Gene Symbols TRIAP1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human TRIAP1. Recombinant protein control fragment (Product #RP-92082).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51499
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.