Learn More
TRIB1 Mouse anti-Human, Clone: OTI1F11, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF805372

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
TRIB1 also called gprotein coupled receptor induced, skip1, gig2, c8fwATP, functions in binding protein binding, protein kinase inhibitor activity, and protein serine/threonine kinase activities. Overexpression of TRIB1 in HeLa cells repressed the basal activity of the IL8 promoter by inhibiting AP1 activity. Overexpression of TRIB1 inhibited oncogenic Ras -driven AP1 activation and MEKK1-mediated AP1 activation. ERK activation was enhanced by TRIB1. Coimmunoprecipitation and yeast 2-hybrid assays showed that MEK1 interacted with both TRIB1 and TRIB3, and MKK4 interacted specifically with TRIB1. Cotransfection of MKK4 enhanced the level of TRIB1, indicating that the TRIB-MAPKK interaction stabilized TRIB1.Specifications
TRIB1 | |
Monoclonal | |
Unconjugated | |
TRIB1 | |
C8FW, GIG-2, GIG2, SKIP1, TRB-1, TRB1 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10221 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI1F11 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q96RU8 | |
TRIB1 | |
Full length human recombit protein of human TRIB1 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.