Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRIB1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA514801
Description
This antibody is predicted to react with mouse based on sequence homology.
TRIB1 also called gprotein coupled receptor induced, skip1, gig2, c8fwATP, functions in binding protein binding, protein kinase inhibitor activity, and protein serine/threonine kinase activities. Overexpression of TRIB1 in HeLa cells repressed the basal activity of the IL8 promoter by inhibiting AP1 activity. Overexpression of TRIB1 inhibited oncogenic Ras -driven AP1 activation and MEKK1-mediated AP1 activation. ERK activation was enhanced by TRIB1. Coimmunoprecipitation and yeast 2-hybrid assays showed that MEK1 interacted with both TRIB1 and TRIB3, and MKK4 interacted specifically with TRIB1. Cotransfection of MKK4 enhanced the level of TRIB1, indicating that the TRIB-MAPKK interaction stabilized TRIB1.
Specifications
| TRIB1 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q96RU8 | |
| TRIB1 | |
| KLH conjugated synthetic peptide between 341-370 amino acids from the C-terminal region of human TRIB1. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| TRIB1 | |
| A530090O15Rik; C8FW; Gig2; GIG-2; G-protein-coupled receptor induced protein; G-protein-coupled receptor induced protein GIG2; G-protein-coupled receptor-induced gene 2 protein; G-protein-coupled receptor-induced protein 2; phosphoprotein regulated by mitogenic pathways; SKIP1; Trb1; TRB-1; Trib1; tribbles homolog 1; tribbles homolog 1 (Drosophila); tribbles pseudokinase 1; tribbles-like protein 1 | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 10221 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction