Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TRIB2 Antibody (OTI8D11), Allophycocyanin, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274597APC
Description
TRIB2 Monoclonal specifically detects TRIB2 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
TRIB2 | |
Monoclonal | |
APC | |
PBS with 0.05% Sodium Azide | |
TRIB2 | |
Full length recombinant protein of human TRIB2 (NP_067675) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse | |
IgG2b |
Flow Cytometry | |
OTI8D11 | |
Flow Cytometry | |
C5FW, FLJ57420, GS3955, TRB-2, TRB2tribbles homolog 2, tribbles homolog 2 (Drosophila) | |
Mouse | |
Affinity Purified | |
Protein Kinase | |
28951.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction