Learn More
Invitrogen™ TRIB3 Monoclonal Antibody (OTI3C6)

Description
TRIB3 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | TRIB3 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI3C6 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | TRIB3 |
| Gene Accession No. | Q96RU7 |
| Gene Alias | C20orf97; DJ1103G7.3; Ifld2; induced in fatty liver dystrophy 2; kinase; Neuronal cell death Inducible Putative Kinase; neuronal cell death-inducible putative kinase; Nipk; p65-interacting inhibitor of NF-kappaB; p65-interacting inhibitor of NF-kappa-B; SINK; SKIP3; Trb3; TRB-3; TRIB3; Tribbles 3; Tribbles homolog 3; tribbles homolog 3 (Drosophila); tribbles pseudokinase 3 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.