Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ TRIB3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA596319
Description
Positive Samples: 293T, SK-HEP1, K-562, SKOV3, BT-474, Mouse liver; Cellular Location: Nucleus Immunogen sequence: MRATPLAAPA GSLSRKKRLE LDDNLDTERP VQKRARSGPQ PRLPPCLLPL SPPTAPDRAT AVATASRLGP YVLLEPEEGG RAYQALHCPT GTEYTCKVYP VQEALAVLEP YARLPPHKHV ARPTEVLAGT.
A novel inhibitor of cell division, Tribbles, blocks mitosis at a critical point in Drosophila morphogenesis. Recently, mammalian homologues of Tribbles (Trbs) have been identified that control mitogen-activated protein kinase (MAPK) activity. TRIB3 also known as TRB-3, SINK, and SKIP3 is a member of Trbs family. Overexpression of this protein inhibits NF-kB-dependent transcription induced by tumor necrosis factor (TNF) stimulation or its downstream signaling proteins but does not inhibit NF-kB translocation to the nucleus and binding to DNA. TRIB3/SKIP3 is also overexpressed in multiple human tumors and is regulated by hypoxia.
Specifications
TRIB3 | |
Polyclonal | |
Unconjugated | |
TRIB3 | |
C20orf97; DJ1103G7.3; Ifld2; induced in fatty liver dystrophy 2; kinase; Neuronal cell death Inducible Putative Kinase; neuronal cell death-inducible putative kinase; Nipk; p65-interacting inhibitor of NF-kappaB; p65-interacting inhibitor of NF-kappa-B; SINK; SKIP3; Trb3; TRB-3; TRIB3; Tribbles 3; Tribbles homolog 3; tribbles homolog 3 (Drosophila); tribbles pseudokinase 3 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
228775, 57761 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
0.55 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K4K2, Q96RU7 | |
TRIB3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human TRIB3 (NP_0669812). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction