Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ TRIB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114501
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114501 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114501 Supplier Invitrogen™ Supplier No. PA5114501
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive controls that may be used include: Mouse Kidney Tissue Lysate.

A novel inhibitor of cell division, Tribbles, blocks mitosis at a critical point in Drosophila morphogenesis. Recently, mammalian homologues of Tribbles (Trbs) have been identified that control mitogen-activated protein kinase (MAPK) activity. TRIB3 also known as TRB-3, SINK, and SKIP3 is a member of Trbs family. Overexpression of this protein inhibits NF-kB-dependent transcription induced by tumor necrosis factor (TNF) stimulation or its downstream signaling proteins but does not inhibit NF-kB translocation to the nucleus and binding to DNA. TRIB3/SKIP3 is also overexpressed in multiple human tumors and is regulated by hypoxia.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TRIB3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TRIB3
Gene Accession No. Q8K4K2, Q96RU7, Q9WTQ6
Gene Alias C20orf97; DJ1103G7.3; Ifld2; induced in fatty liver dystrophy 2; kinase; Neuronal cell death Inducible Putative Kinase; neuronal cell death-inducible putative kinase; Nipk; p65-interacting inhibitor of NF-kappaB; p65-interacting inhibitor of NF-kappa-B; SINK; SKIP3; Trb3; TRB-3; TRIB3; Tribbles 3; Tribbles homolog 3; tribbles homolog 3 (Drosophila); tribbles pseudokinase 3
Gene Symbols TRIB3
Host Species Rabbit
Immunogen TRB3 antibody was raised against a 17 amino acid peptide near the center of human TRB3. The immunogen is located within amino acids 100 - 150 of TRB3.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 228775, 246273, 57761
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.