Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tribenzylsilane 98.0+%, TCI America™
SDP

Catalog No. T191625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T191625G 25 g
T19165G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T191625G Supplier TCI America Supplier No. T191625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1747-92-8
Molecular Formula C21H21Si
Molecular Weight (g/mol) 301.48
MDL Number MFCD00004769
InChI Key QXZGXRIXJAVMTI-UHFFFAOYSA-N
PubChem CID 6330251
IUPAC Name tribenzylsilyl
SMILES C([Si](CC1=CC=CC=C1)CC1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 90°C
Color White-Yellow
Boiling Point 175°C
Quantity 25 g
Formula Weight 302.49
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Tribenzylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.