Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyl Borate 99.0+%, TCI America™
SDP

Catalog No. B0518100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B0518100ML 100 mL
B051825ML 25 mL
B0518500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B0518100ML Supplier TCI America Supplier No. B0518100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 688-74-4
Molecular Formula C12H27BO3
Molecular Weight (g/mol) 230.155
MDL Number MFCD00009434
InChI Key LGQXXHMEBUOXRP-UHFFFAOYSA-N
Synonym n-butyl borate, tri-n-butyl borate, tributoxyborane, butyl borate, tributoxyboron, boron tributoxide, borester 2, tri-n-butoxyborane, tris butoxy borane, borane, tributoxy
PubChem CID 12712
IUPAC Name tributyl borate
SMILES B(OCCCC)(OCCCC)OCCCC

Specifications

Color Colorless
Boiling Point 230°C
Quantity 100 mL
Formula Weight 230.16
Percent Purity ≥99.0% (T)
Physical Form Liquid
Chemical Name or Material Tributyl Borate
EINECSNumber (2)-2101
RTECSNumber ED4900000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.