Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyl(vinyl)tin, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
7486-35-3
C14H30Sn
317.09
MFCD00009421
QIWRFOJWQSSRJZ-UHFFFAOYSA-N
tributyl vinyl tin,vinyltributyltin,vinyltributylstannane,tributyl vinyl stannane,tributyl ethenyl stannane,stannane, tributylethenyl,vinyltributyl tin,stannane, tributylvinyl,tributylstannylethylene,vinyltri-n-butyltin
81998
tributyl(ethenyl)stannane
CCCC[Sn](CCCC)(CCCC)C=C
7486-35-3
C14H30Sn
317.09
MFCD00009421
QIWRFOJWQSSRJZ-UHFFFAOYSA-N
tributyl vinyl tin,vinyltributyltin,vinyltributylstannane,tributyl vinyl stannane,tributyl ethenyl stannane,stannane, tributylethenyl,vinyltributyl tin,stannane, tributylvinyl,tributylstannylethylene,vinyltri-n-butyltin
81998
tributyl(ethenyl)stannane
CCCC[Sn](CCCC)(CCCC)C=C
Flash Point >110°C
Boiling Point 104°C to 106°C (3.5mmHg)
Refractive Index 1.4770 to 1.479
Specific Gravity 1.08
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 25g
Color Colorless to Yellow
Density 1.0800g/mL
Assay Percent Range 96% min. (GC)
Solubility Information Solubility in water: .. Other solubilities: soluble in all the common organic solvents
Formula Weight 317.09
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Tributyl(vinyl)tin
Show More Show Less

GHS H Statement:
Causes damage to organs through prolonged or repeated exposure.
Harmful in contact with skin.
Causes skin irritation.
Toxic if swallowed.
Very toxic to aquatic life with long lasting effects.
Causes serious eye irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 231-291-4