Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributylchlorosilane 97.0+%, TCI America™
SDP

Catalog No. T352425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T352425G 25 g
T35245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T352425G Supplier TCI America Supplier No. T352425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 995-45-9
Molecular Formula C12H27ClSi
Molecular Weight (g/mol) 234.883
MDL Number MFCD00000509
InChI Key JSQJUDVTRRCSRU-UHFFFAOYSA-N
Synonym silane, tributylchloro, chlorotributylsilane, chlorotri-n-butylsilane, tributyl chloro silane, tri-n-butylchlorosilane, tributylsilyl chloride, tributylchloro-silan, chloro-tri-n-butylsilane
PubChem CID 70448
IUPAC Name tributyl(chloro)silane
SMILES CCCC[Si](CCCC)(CCCC)Cl

Specifications

Color Yellow
Boiling Point 94°C
Quantity 25 g
UN Number 2987
Formula Weight 234.88
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tributylchlorosilane
EINECSNumber (2)-2041
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.