Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tributyltin trifluoromethanesulfonate, 96%

Catalog No. p-3523609
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle

CAS: 68725-14-4 | C13H27F3O3SSn | 439.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68725-14-4
Molecular Formula C13H27F3O3SSn
Molecular Weight (g/mol) 439.13
MDL Number MFCD00043324

Specifications

Melting Point 31°C
Color White to Beige
Boiling Point 56°C to 60°C (25.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Quantity 1 g
Formula Weight 439.13
Percent Purity 96%
Physical Form Crystals and/or Chunks
Chemical Name or Material Tributyltin trifluoromethanesulfonate
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes damage to organs through prolonged or repeated exposure. Causes serious eye damage. Causes severe skin burns and eye damage. Very toxic to aquatic life with long lasting effects. Toxic if swallowed. Harmful in contact with skin. Very toxic to aquatic life.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Do not breathe dust/fume/gas/mist/vapors/spray. Avoid release to the environment.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.