Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tricarballylic Acid 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
50g
99-14-9
C6H8O6
176.13
KQTIIICEAUMSDG-UHFFFAOYSA-N
tricarballylic acid,1,2,3-propanetricarboxylic acid,carballylic acid,tricarballylate,beta-carboxyglutaric acid,propane 1,2,3-tricarboxylic acid,unii-ra5qh2j020,3-carboxyglutaric acid,.beta.-carboxyglutaric acid,1,2,3-propanetricarboxylicacid
14925
CHEBI:45969
propane-1,2,3-tricarboxylic acid
C(C(CC(=O)O)C(=O)O)C(=O)O
99-14-9
C6H8O6
176.13
KQTIIICEAUMSDG-UHFFFAOYSA-N
tricarballylic acid,1,2,3-propanetricarboxylic acid,carballylic acid,tricarballylate,beta-carboxyglutaric acid,propane 1,2,3-tricarboxylic acid,unii-ra5qh2j020,3-carboxyglutaric acid,.beta.-carboxyglutaric acid,1,2,3-propanetricarboxylicacid
14925
CHEBI:45969
propane-1,2,3-tricarboxylic acid
C(C(CC(=O)O)C(=O)O)C(=O)O
Infrared Spectrum Authentic
Packaging Glass bottle
Quantity 50g
Color White to Brown
Melting Point 154°C to 159°C
Assay Percent Range 99%
Linear Formula HO2CCH2CH(CO2H)CH2CO2H
Beilstein 02, 815
Merck Index 15, 9787
Formula Weight 176.13
Physical Form Fine Crystalline Powder
Percent Purity ≥98.5%
Chemical Name or Material Tricarballylic acid, 99%
Show More Show Less

GHS P Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 202-733-3

TSCA : TSCA