Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichlormethiazide, USP, 98-102%, Spectrum™ Chemical
SDP

Catalog No. 18607540 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
1 kg
5 kg
Packaging:
Amber Glass Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-540 100 g Amber Glass Bottle
18-607-538 5 g Amber Glass Bottle
18-607-539 25 g Amber Glass Bottle
18-607-541 1 kg Amber Glass Bottle
18-607-542 5 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-607-540 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T1435100GM

T1435, 133-67-5, C8H8Cl3N3O4S2

Spectrum™ Chemical Trichlormethiazide, USP is a thiazide diuretic and is used to treat edema and high blood pressure. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 133-67-5
Molecular Formula C8H8Cl3N3O4S2
Molecular Weight (g/mol) 380.64
InChI Key LMJSLTNSBFUCMU-UHFFFAOYNA-N
IUPAC Name 6-chloro-3-(dichloromethyl)-1,1-dioxo-3,4-dihydro-2H-1λ⁶,2,4-benzothiadiazine-7-sulfonamide
SMILES NS(=O)(=O)C1=CC2=C(NC(NS2(=O)=O)C(Cl)Cl)C=C1Cl

Specifications

CAS Max % 1
Ignition Residue 0.001
Loss on Drying 0.005
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 98 to 102%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.