Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetic Acid, 100 % (w/v) Aqueous Solution, Spectrum™ Chemical
SDP

Catalog No. 18607544
Click to view available options
Quantity:
1 L
20 L
Packaging:
Amber Glass Bottle
Poly Drum

T-184, 76-03-9

Spectrum™ Chemical Trichloroacetic Acid, 100 Percent (w/v) Aqueous Solution , also known as TCA, is used in biochemistry to help the precipitation of macromolecules. It is also used in the cosmetic industry as a chemical peel and also as a wart remover.

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.38
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
IUPAC Name trichloroacetic acid
SMILES OC(=O)C(Cl)(Cl)Cl

Specifications

CAS Max % 1
Percent Purity 100% ±2%
Grade Chemical Solution
Quantity 1 L
Packaging Amber Glass Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.