Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
                
Learn More
Learn More
        Trichloroacetic Acid, 33% (w/v), Ricca Chemical
    
    
    
        
            Shop All Ricca Chemical Company Products
        
    
    
    
    
    
    
    
    
        
                    Click to view available options
                
                
                    
                            
                                Quantity:
                            
                        
                        
                            
                                120 mL
                            
                        
                            
                                500 mL
                            
                        
                            
                                1 L
                            
                        
                            
                                4 L
                            
                        
                    
                            
                                Packaging:
                            
                        
                        
                            
                                Amber Glass Bottle
                            
                        
                    Specifications
Specifications
| CAS Min % | 24.29, 74.0, 0.15 | 
| Color | Colorless | 
| CAS Max % | 25.28, 77.0, 0.15 | 
| Percent Purity | 33% w/v | 
| Grade | Laboratory | 
| Concentration or Composition (by Analyte or Components) | 33% (w/v) Aqueous Solution | 
| Quantity | 1 L | 
| Packaging | Amber Glass Bottle | 
| Physical Form | Liquid | 
| Chemical Name or Material | Trichloroacetic Acid | 
Safety and Handling
| ShelfLife | 12 months | 
                    Product Content Correction
                
                Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
			
            Spot an opportunity for improvement?Share a Content Correction