Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetic Acid, 5% w/v Solution, PROMO, Thermo Scientific Chemicals

Catalog No. J62366K2PM
Click to view available options
Quantity:
1 L

Protein precipitation reagent

Applications

Trichloroacetic acid, is traditionally used to precipitate protein. Has been used to determine protein concentration by quantitative precipitation. It is also used as a decalcifier and fixative in microscopy in protein sequencing, detecting albumin and organic synthesis. A protocol for the precipitation of nucleic acids can be found in Molecular Cloning.

Solubility

Soluble in water (1200 mg/mL at 20°C), alcohol, ether, ethanol (very soluble), and carbon tetrachloride.

Notes

Store under nitrogen. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents, strong bases, smines.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.38 g/mol
MDL Number MFCD00004177
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym trichloroacetic acid, trichloroethanoic acid, aceto-caustin, trichloroacetic acid, acetic acid, trichloro, trichloressigsaeure, konesta, acide trichloracetique, acido tricloroacetico, amchem grass killer
PubChem CID 6421
ChEBI CHEBI:30956
IUPAC Name 2,2,2-trichloroacetic acid
SMILES C(=O)(C(Cl)(Cl)Cl)O

Specifications

Color Colorless
UN Number UN2564
Beilstein 970119
Merck Index 14,9627
Solubility Information Soluble in water, alcohol, ether, ethanol, and carbon tetrachloride.
Concentration 5% w/v
Quantity 1 L
Physical Form Liquid
Chemical Name or Material Trichloroacetic acid
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: TRICHLOROACETIC ACID, SOLUTION
EINECSNumber 200-927-2
TSCA Yes

RUO - Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.