Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetic Acid, 85% (w/v) Aqueous Solution, Spectrum™ Chemical
SDP

Catalog No. 18614519 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-519 1 L Amber Glass Bottle
18-614-518 500 mL Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-614-519 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T1831LTGL

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

Chemical Identifiers

CAS 76-03-9 , 7732-18-5

Specifications

Percent Purity 85% ±2.0%
Grade Chemical Solution
Quantity 1 L
Packaging Amber Glass Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.