Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetic acid Solution, MP Biomedicals™

Catalog No. MP219605790 Shop All MP Biomedicals, Inc Products
Click to view available options
Quantity:
500 mL

Trichloroacetic acid Solution

  • It is used in protein precipitation; has been used to determine protein concentration by quantitative precipitation.
  • It is also Used as a decalcifier and fixative in microscopy.
  • A protocol for the precipitation of nucleic acids can be found in Molecular Cloning. Rats exposed to chronic sublethal amounts of trichloroacetic acid displayed an increase in serum bilirubin with a decrease in hematological proteins and cholesterols along with significant decreases in red blood corpuscles, mean cell volume, mean corpuscular hemoglobin, hemoglobin and hematocrit.
  • In mice the production of liver tumors was associated to Trichloroacetic acid, which is also a known mouse hepatocarinogen.
  • On the guinea pig trichloroacetic acid was observed to be a mild alergen on the skin.
  • Trichloroacetic acid is an acetic acid analogue commonly used to precipitate proteins, DNA and RNA. In the presence of SDS Trichloroacetic acid will precipitate proteins which can then be quantified by the Lowry method.
  • The compound has also found uses as a decalcifier and fixative in microscopy, in protein sequencing, detecting albumin and organic synthesis.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 76-03-9
Molecular Formula C2HCl3O2
Molecular Weight (g/mol) 163.38
MDL Number MFCD00004177
InChI Key YNJBWRMUSHSURL-UHFFFAOYSA-N
Synonym Trichloroethanoic Acid, TCA, Aceto-caustin, Acide trichloracetique

Specifications

pH 1.2 (0.1 M aqueous solution)(literature)
Melting Point ∼56°C (literature)
Boiling Point 197°C at 1013 hPa(Literature)
UN Number UN2564
Beilstein 970119
Density 1.61 g/cm3 (Lit.)
Quantity 500 mL
Physical Form Solution
Vapor Density 5.6 (air = 1)(Lit.)
Vapor Pressure 1.2 hPa at 50°C(Lit.)
RTECSNumber AJ7875000
storageNote1 +4°C
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.