Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroacetic anhydride, 95%

Catalog No. p-4440188
Click to view available options
Quantity:
25 g
100 g

CAS: 4124-31-6 | C4Cl6O3 | 308.76 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4124-31-6
Molecular Formula C4Cl6O3
Molecular Weight (g/mol) 308.76
InChI Key MEFKFJOEVLUFAY-UHFFFAOYSA-N
Synonym trichloroacetic anhydride, acetic acid, trichloro-, anhydride, acetic acid, 2,2,2-trichloro-, 1,1'-anhydride, acmc-1aliy, perchloroacetic anhydride, bis trichloroacetic anhydride, trichloroacetic acid anhydride, 2,2,2-trichloroacetic anhydride, trichloroacetyl 2,2,2-trichloroacetate
PubChem CID 20079
IUPAC Name (2,2,2-trichloroacetyl) 2,2,2-trichloroacetate
SMILES C(=O)(C(Cl)(Cl)Cl)OC(=O)C(Cl)(Cl)Cl

Specifications

Density 1.6900g/mL
Boiling Point 139°C to 141°C (60.0 mmHg)
Packaging Glass Bottle
Linear Formula [CCl3C(=O)]2O
Quantity 25 g
Specific Gravity 1.69
Solubility Information Solubility in water: reacts
Formula Weight 308.76
Percent Purity 95%
Chemical Name or Material Trichloroacetic anhydride
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
EINECSNumber 223-925-3

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.