Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloroethylene, ACS, 99.5%, Spectrum™ Chemical
SDP

Catalog No. 18614524 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-524 4 L Amber Glass Bottle
18-614-527 500 mL Amber Glass Bottle
18-614-525 20 L Steel Can
18-614-526 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Catalog No. 18-614-524 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T11154LTGL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Chemical Identifiers

CAS 79-01-6
Molecular Formula C2HCl3
Molecular Weight (g/mol) 131.38
InChI Key XSTXAVWGXDQKEL-UHFFFAOYSA-N
IUPAC Name 1,1,2-trichloroethene
SMILES ClC=C(Cl)Cl

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 4 L
Residue after Evaporation 1e-05
Percent Purity 0.995
Grade ACS
Titratable Acid 0.0001 meq/g
WARNING: Cancer - www.P65Warnings.ca.gov

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.