Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Trichloromethiazide, MP Biomedicals™

Catalog No. ICN19002001 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19002001 1 g
ICN19002010 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN19002001 Supplier MP Biomedicals, Inc Supplier No. 0219002001

Chemical Identifiers

CAS 133-67-5
Molecular Formula C8H8Cl3N3O4S2
Molecular Weight (g/mol) 380.639
MDL Number MFCD00057315
InChI Key LMJSLTNSBFUCMU-UHFFFAOYSA-N
Synonym trichlormethiazide, trichloromethiazide, naqua, trichlormetazid, metahydrin, achletin, diurese, trichlormethiazid, diu-hydrin, chlopolidine
PubChem CID 5560
ChEBI CHEBI:9683
IUPAC Name 6-chloro-3-(dichloromethyl)-1,1-dioxo-3,4-dihydro-2H-1$l^{6},2,4-benzothiadiazine-7-sulfonamide
SMILES C1=C2C(=CC(=C1Cl)S(=O)(=O)N)S(=O)(=O)NC(N2)C(Cl)Cl

Specifications

Color White
Quantity 1 g
Formula Weight 380.67
Physical Form Powder
Chemical Name or Material Trichloromethiazide
EINECSNumber 205-118-8
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.