Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Trichomonas Vaginalis 1 Antibody (1306/156), Alexa Fluor™ 488, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NB10062964AF488
Description
Trichomonas Vaginalis 1 Monoclonal specifically detects Trichomonas Vaginalis 1 in Protozoa samples. It is validated for ELISA.Specifications
Trichomonas Vaginalis 1 | |
Monoclonal | |
Alexa Fluor 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
Mouse | |
Protein A purified | |
RUO | |
Recognises T. vaginalis strain 1, an anaerobic parasitic protozoan and causative agent of the sexually transmitted disease Trichomoniasis. | |
Store at 4C in the dark. | |
IgG1 |
ELISA | |
1306/156 | |
ELISA | |
T vaginalis, TV | |
Native | |
0.1 mL | |
Primary | |
Protozoa | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction