Learn More
Invitrogen™ Trichomonas vaginalis Monoclonal Antibody (7101)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA547579
Description
Specificity: Specific for T. vaginalis. Best ELISA pairs (capture/conjugate): Trichomonas vaginalis Monoclonal antibody (Product # MA5-18275)/Trichomonas vaginalis Monoclonal antibody (Product # MA5-47579); Trichomonas vaginalis Monoclonal antibody (Product # MA5-47580)/Trichomonas vaginalis Monoclonal antibody (Product # MA5-47579). Purified preparations consist of >90% pure mouse monoclonal antibody which has been purified from ascites fluid by protein A chromatography. The final preparation is formulated to a protein concentration of 100 μg/mL in 0.01 M phosphate buffered saline, pH 7.2 and contains 0.1% sodium azide. Each vial contains 1.0 mL. This product contains no stabilizing proteins and should be stored at 4C until ready for use. Working dilution must be determined by the user. Suggested starting ranges are 1:10-1:50 for ICC/IF and 1:20-1:200. for ELISA.

Specifications
Trichomonas vaginalis | |
Monoclonal | |
100 μg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Mouse | |
Protein A | |
RUO | |
Protozoa | |
Antibody | |
IgG3 |
ELISA, Immunocytochemistry | |
7101 | |
Unconjugated | |
t vaginalis; T. vaginalis | |
Proprietary sequence. | |
100 μg | |
Primary | |
4°C | |
Liquid |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.