Learn More
Trichomonas vaginalis Monoclonal Antibody (A63A)

Description
Trichomonas vaginalis Monoclonal Antibody for ICC/IF
Specifications
Specifications
| Antigen | Trichomonas vaginalis |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | A63A |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | t vaginalis; T. vaginalis |
| Host Species | Mouse |
| Immunogen | Trichomonas vaginalis native protein. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.