Learn More
Invitrogen™ Trichomonas vaginalis Monoclonal Antibody (J37A)

Description
Purity is >90%. Recommended pairs for ELISA: MA5-18274 (capture) and MA5-18273 (conjugated, detection).
Specifications
Specifications
| Antigen | Trichomonas vaginalis |
| Applications | ELISA, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | J37A |
| Concentration | 100 μg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.1% sodium azide; pH 7.2 |
| Gene Alias | t vaginalis; T. vaginalis |
| Host Species | Mouse |
| Immunogen | Trichomonas vaginalis native protein. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.