Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Trichostatin A
GREENER_CHOICESDP

Catalog No. 14061 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-061 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 14-061 Supplier Tocris Bioscience™ Supplier No. 1406/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Potent histone deacetylase inhibitor

Trichostatin A is a selective and potent inhibitor of class I and II histone deacetylases (HDAC) (Ki = 3.4 nM, IC50 values are 4.99 nM, 5.21 nM, 16.4 nM, 24.3 nM, 27.6 nM for HDAC1, 3, 6, 10 and 4, respectively). Trichostatin A induces accelerated dedifferentiation of primordial germ cells (PGCs) into embryonic germ (EG) cells. Trichostatin A potently inhibits a range of breast cancer and melanoma cell lines (IC50 = 29-400 nM) and induces apoptosis in prostate cancer cells. In an animal model of Huntington's disease, inhibition of HDAC by Trichostatin A rescues memory deficits and increases expression of CREB/CBP target genes in mutant mice. Activates autophagy, increases vesicular transport of BDNF (Cat. No. 2837) and is active in vivo.

Chemical Identifiers

CAS 58880-19-6
Molecular Formula C17H22N2O3

Specifications

Quantity 1 mg
Formula Weight 302.37
Purity 97%
Molecular Weight (g/mol) 302.37
Chemical Name or Material Trichostatin A
Recommended Storage Desiccate at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.