Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Triclabendazole, 98%

Catalog No. AC461960010
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461960010 1 g
AC461960050 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461960010 Supplier Thermo Scientific Chemicals Supplier No. 461960010
Add to Cart

CAS: 68786-66-3 | C14H9Cl3N2OS | 359.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68786-66-3
Molecular Formula C14H9Cl3N2OS
Molecular Weight (g/mol) 359.66
InChI Key NQPDXQQQCQDHHW-UHFFFAOYSA-N
Synonym triclabendazole, fasinex, 6-chloro-5-2,3-dichlorophenoxy-2-methylthio-1h-benzo d imidazole, triclabendazol inn-spanish, triclabendazolum inn-latin, ccris 8988, 5-chloro-6-2,3-dichlorophenoxy-2-methylthio-1h-benzimidazole, 6-chloro-5-2,3-dichlorophenoxy-2-methylthio-benzimidazole, 5-chloro-6-2,3-dichlorophenoxy-2-methylthio benzimidazole, 6-chloro-5-2,3-dichlorophenoxy-2-methylthio-1h-benzimidazole
PubChem CID 50248
IUPAC Name 6-chloro-5-(2,3-dichlorophenoxy)-2-methylsulfanyl-1H-benzimidazole
SMILES CSC1=NC2=CC(=C(C=C2N1)Cl)OC3=C(C(=CC=C3)Cl)Cl

Specifications

Melting Point 175.0°C to 176.0°C
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble.
Formula Weight 359.66
Percent Purity 98%
Chemical Name or Material Triclabendazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.