Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tridecafluorohexyl Bromide 97.0+%, TCI America™
SDP

Catalog No. T247925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T247925G 25 g
T24795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T247925G Supplier TCI America Supplier No. T247925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 335-56-8
Molecular Formula C6BrF13
Molecular Weight (g/mol) 398.949
MDL Number MFCD00042349
InChI Key JTYRBFORUCBNHJ-UHFFFAOYSA-N
Synonym 1-bromoperfluorohexane, perfluorohexyl bromide, perfluorohexylbromide, tridecafluorohexyl bromide, 1-bromo-tridecafluorohexane, bromoperfluorohexane, acmc-1cmhf, 1-bromotridecafluorohexane, tridecafluoro-1-bromohexane
PubChem CID 92755
IUPAC Name 1-bromo-1,1,2,2,3,3,4,4,5,5,6,6,6-tridecafluorohexane
SMILES C(C(C(C(F)(F)Br)(F)F)(F)F)(C(C(F)(F)F)(F)F)(F)F

Specifications

Color Yellow
Boiling Point 100°C
Quantity 25 g
Formula Weight 398.95
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Tridecafluorohexyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.