Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tridecyl Methacrylate (mixture of branched chain isomers) (stabilized with MEHQ), TCI America™
SDP

Catalog No. M026425ML
Click to view available options
Quantity:
25 mL

Chemical Identifiers

CAS 2495-25-2
Molecular Formula C17H32O2
Molecular Weight (g/mol) 268.441
MDL Number MFCD00059220
InChI Key KEROTHRUZYBWCY-UHFFFAOYSA-N
Synonym Methacrylic Acid Tridecyl Ester
PubChem CID 17234
IUPAC Name tridecyl 2-methylprop-2-enoate
SMILES CCCCCCCCCCCCCOC(=O)C(=C)C

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 268.44
Physical Form Liquid
Chemical Name or Material Tridecyl Methacrylate (mixture of branched chain isomers) (stabilized with MEHQ)
EINECSNumber (2)-1039
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.