Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethanolamine, BAKER ANALYZED™ Reagent, J.T. Baker™

Catalog No. 02004533
Click to view available options
Quantity:
500 mL
4 L
Packaging:
Narrow Mouth Amber Glass Bottle

High quality chemicals for laboratory and specialized industrial use

Chemical Identifiers

CAS 102-71-6
Molecular Formula C6H15NO3
Molecular Weight (g/mol) 149.19
InChI Key GSEJCLTVZPLZKY-UHFFFAOYSA-N
Synonym triethanolamine, trolamine, 2,2',2-nitrilotriethanol, sterolamide, daltogen, tris 2-hydroxyethyl amine, triethylolamine, nitrilotriethanol, trihydroxytriethylamine, thiofaco t-35
PubChem CID 7618
ChEBI CHEBI:28621
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethanol
SMILES C(CO)N(CCO)CCO

Specifications

Flash Point 179.00°C
Packaging Narrow Mouth Amber Glass Bottle
Quantity 4 L
Specific Gravity 1L = 1.13kg
Formula Weight 149.19
Grade BAKER ANALYZED™ Reagent
Chemical Name or Material Triethanolamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.