Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethanolamine Salicylate, 85-95%, Spectrum™ Chemical
SDP

Catalog No. 18614540 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
10 kg
250 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-614-540 10 kg Amber Glass Bottle
18-614-539 100 g Amber Glass Bottle
18-614-541 500 g Amber Glass Bottle
18-614-536 1 kg Amber Glass Bottle
18-614-537 250 kg Amber Glass Bottle
18-614-538 25 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-614-540 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T111410KGBL
Add to Cart
Add to Cart
Request a Quote

Chemical Identifiers

CAS 2174-16-5

Specifications

CAS Max % 1
pH 6.0to 7.0
Packaging Amber Glass Bottle
Quantity 10 kg
Molecular Weight (g/mol) 287.18
Percent Purity 85 to 95%
Grade Reagent
Physical Form Liquid
Chemical Name or Material Triethanolamine Salicylate

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.