Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Triethoxyvinylsilane

Catalog No. 175560500ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
100 mL
500 mL
Catalog No. Quantity
17-556-0500ML 500 mL
17-556-0100ML 100 mL
2 options

Catalog No. 17-556-0500ML

Supplier: Sigma Aldrich 175560500ML

Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Triethoxyvinylsilane(TEVS) is a silane coupling agent and an adhesion promoter which can be used for cross-linking and for providing an insulating layer with good mechanical and thermal properties.Triethoxyvinylsilane (TEVS) was used to modify nano-silica particlesand poly (vinyl acetate). TEVS may undergo emulsion copolymerization with styrene (St), butyl acrylate (BA), and methacrylic acid (MAA). The formation of random copolymers of TEVS with styrene (St) has been reported. TEVS may undergo graft polymerization with styrene and methyl methacrylate over different silicates

Specifications

CAS 78-08-0
Density 0.903 g/mL (at 25°C)
Boiling Point 160°C to 161°C; 62°C to 63°C (20 mmHg)
Molecular Formula C8H18O3Si
Refractive Index n20/D 1.398 (literature)
Linear Formula H2C=CHSi(OC2H5)3
Quantity 500 mL
MDL Number MFCD00009063
Synonym (Triethoxysilyl)ethylene; Vinyltriethoxysilane
Molecular Weight (g/mol) 190.31
Percent Purity 97%
Biological Activity Respiratory system
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.