Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethoxyvinylsilane, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
500g
Packaging:
Glass bottle
78-08-0
C8H18O3Si
190.31
MFCD00009063
FWDBOZPQNFPOLF-UHFFFAOYSA-N
vinyltriethoxysilane, triethoxyvinylsilane, silane, ethenyltriethoxy, triethoxy vinyl silane, polyscience vtes, triethoxyvinylsilicane, vtes, silane a 151, ethenyl triethoxy silane, silane, triethoxyvinyl
6516
ethenyltriethoxysilane
CCO[Si](OCC)(OCC)C=C
78-08-0
C8H18O3Si
190.31
MFCD00009063
FWDBOZPQNFPOLF-UHFFFAOYSA-N
vinyltriethoxysilane, triethoxyvinylsilane, silane, ethenyltriethoxy, triethoxy vinyl silane, polyscience vtes, triethoxyvinylsilicane, vtes, silane a 151, ethenyl triethoxy silane, silane, triethoxyvinyl
6516
ethenyltriethoxysilane
CCO[Si](OCC)(OCC)C=C
Flash Point 34°C
Boiling Point 160°C to 161°C
Specific Gravity 0.9
Refractive Index 1.3970 to 1.399
Quantity 25g
Infrared Spectrum Authentic
Packaging Glass bottle
Density 0.9000g/mL
Color Colorless
Assay Percent Range 96% min. (GC)
Linear Formula H2C=CHSi(OC2H5)3
Solubility Information Solubility in water: decomposes in water. Other solubilities: soluble in chloroform
Formula Weight 190.32
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material Triethoxyvinylsilane, 97%
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 201-081-7

RTECSNumber : VV6700000

TSCA : TSCA