Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl 1,3,5-benzenetricarboxylate, 97%

Catalog No. p-7026980
Click to view available options
Quantity:
5 g
25 g

CAS: 4105-92-4 | C15H18O6 | 294.303 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4105-92-4
Molecular Formula C15H18O6
Molecular Weight (g/mol) 294.303
MDL Number MFCD00236389
InChI Key KXGOWZRHSOJOLF-UHFFFAOYSA-N
Synonym triethyl trimesate, 1,3,5-benzenetricarboxylic acid, triethyl ester, trimesic acid triethyl ester, triethyl 1,3,5-benzenetricarboxylate, ethyl 3,5-bis ethoxycarbonyl benzoate, 1,3,5-triethyl benzene-1,3,5-tricarboxylate, acmc-20a6fr, bzta02, 1,3,5-tri ethoxycarbonyl benzene, triethyl 1,5-benzenetricarboxylate
PubChem CID 226764
IUPAC Name triethyl benzene-1,3,5-tricarboxylate
SMILES CCOC(=O)C1=CC(=CC(=C1)C(=O)OCC)C(=O)OCC

Specifications

Melting Point 132°C to 135°C
Quantity 5 g
Formula Weight 294.31
Percent Purity 97%
Chemical Name or Material Triethyl 1,3,5-benzenetricarboxylate
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.