Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl 2-phosphonopropionate, 98%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
3699-66-9
C9H19O5P
238.22
MFCD00009159
BVSRWCMAJISCTD-UHFFFAOYSA-N
triethyl 2-phosphonopropionate,ethyl 2-diethoxyphosphoryl propanoate,ethyl-2-diethylphosphono propanoate,diethyl 1-carbethoxy ethylphosphonate,propanoic acid, 2-diethoxyphosphinyl-, ethyl ester,diethyl ethoxycarbonylethyl-phosphonate,diethyl 1-ethoxycarbonyl ethanephosphonate,2-diethoxyphosphinyl propanoic acid ethyl ester,diethyl 1-ethoxycarbonyl ethanephosphonate~2-phosphonopropionic acid triethyl ester,triethyl2-phosphonopropionate
107155
ethyl 2-diethoxyphosphorylpropanoate
CCOC(=O)C(C)P(=O)(OCC)OCC
3699-66-9
C9H19O5P
238.22
MFCD00009159
BVSRWCMAJISCTD-UHFFFAOYSA-N
triethyl 2-phosphonopropionate,ethyl 2-diethoxyphosphoryl propanoate,ethyl-2-diethylphosphono propanoate,diethyl 1-carbethoxy ethylphosphonate,propanoic acid, 2-diethoxyphosphinyl-, ethyl ester,diethyl ethoxycarbonylethyl-phosphonate,diethyl 1-ethoxycarbonyl ethanephosphonate,2-diethoxyphosphinyl propanoic acid ethyl ester,diethyl 1-ethoxycarbonyl ethanephosphonate~2-phosphonopropionic acid triethyl ester,triethyl2-phosphonopropionate
107155
ethyl 2-diethoxyphosphorylpropanoate
CCOC(=O)C(C)P(=O)(OCC)OCC
For C-C Bond Formation (Olefination)
Boiling Point 143.0°C to 144.0°C (12.0mmHg)
Specific Gravity 1.11
Packaging Glass bottle
Infrared Spectrum Authentic
Density 1.1100g/mL
Quantity 50g
Flash Point 88°C
Refractive Index 1.4310 to 1.4330
Color Colorless
Assay Percent Range 98%
Linear Formula (C2H5O)2P(O)CH(CH3)CO2C2H5
Beilstein 04, I, 573
Formula Weight 238.22
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Triethyl 2-phosphonopropionate, 98%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautious

GHS Signal Word: Warning

EINECSNumber : 223-033-4