Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl citrate, 99%

Catalog No. AC375060010
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle

CAS: 77-93-0 | C12H20O7 | 276.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77-93-0
Molecular Formula C12H20O7
Molecular Weight (g/mol) 276.29
InChI Key DOOTYTYQINUNNV-UHFFFAOYSA-N
Synonym triethyl citrate, ethyl citrate, citroflex 2, eudraflex, hydragen cat, citric acid, triethyl ester, triaethylcitrat, triethylcitrate, citric acid triethyl ester, 1,2,3-propanetricarboxylic acid, 2-hydroxy-, triethyl ester
PubChem CID 6506
IUPAC Name triethyl 2-hydroxypropane-1,2,3-tricarboxylate
SMILES CCOC(=O)CC(CC(=O)OCC)(C(=O)OCC)O

Specifications

Melting Point -46.0°C
Density 1.1360g/mL
Boiling Point 294.0°C
Flash Point 151°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Refractive Index 1.4400 to 1.4430
Quantity 1 kg
Merck Index 15,2325
Specific Gravity 1.136
Solubility Information Solubility in water: 5.7% (25°C)
Viscosity 35.2 mPa.s (25°C)
Formula Weight 276.29
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Triethyl citrate
Show More Show Less
EINECSNumber 201-070-7

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.