Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl citrate, 99%, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
2.5kg
Packaging:
Plastic bottle
77-93-0
C12H20O7
276.29
DOOTYTYQINUNNV-UHFFFAOYSA-N
triethyl citrate,ethyl citrate,citroflex 2,eudraflex,hydragen cat,citric acid, triethyl ester,triaethylcitrat,triethylcitrate,citric acid triethyl ester,1,2,3-propanetricarboxylic acid, 2-hydroxy-, triethyl ester
6506
triethyl 2-hydroxypropane-1,2,3-tricarboxylate
CCOC(=O)CC(CC(=O)OCC)(C(=O)OCC)O
77-93-0
C12H20O7
276.29
DOOTYTYQINUNNV-UHFFFAOYSA-N
triethyl citrate,ethyl citrate,citroflex 2,eudraflex,hydragen cat,citric acid, triethyl ester,triaethylcitrat,triethylcitrate,citric acid triethyl ester,1,2,3-propanetricarboxylic acid, 2-hydroxy-, triethyl ester
6506
triethyl 2-hydroxypropane-1,2,3-tricarboxylate
CCOC(=O)CC(CC(=O)OCC)(C(=O)OCC)O
Viscosity 35.2 mPa.s (25°C)
Specific Gravity 1.136
Refractive Index 1.4400 to 1.4430
Density 1.1360g/mL
Infrared Spectrum Authentic
Packaging Plastic bottle
Quantity 250g
Boiling Point 294.0°C
Color Undesignated
Flash Point 151°C
Melting Point -46.0°C
Assay Percent Range 98.5% min. (GC)
Merck Index 15,2325
Solubility Information Solubility in water: 5.7% (25°C)
Formula Weight 276.29
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Triethyl citrate
Show More Show Less

EINECSNumber : 201-070-7