Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl Citrate FCC, Reagents

Catalog No. 01336860
Change view
Click to view available options
:
Quantity: 1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-336-860 Quantity: 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-336-860 Supplier Reagents Holdings Llc Supplier No. C33426001KG
Only null left
Add to Cart
Add to Cart

Triethyl Citrate FCC, Meets FCC specifications, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 77-93-0
Molecular Formula C12H20O7
Molecular Weight (g/mol) 276.29
InChI Key DOOTYTYQINUNNV-UHFFFAOYSA-N
IUPAC Name 1,2,3-triethyl 2-hydroxypropane-1,2,3-tricarboxylate
SMILES CCOC(=O)CC(O)(CC(=O)OCC)C(=O)OCC

Specifications

Color Light Yellow
Packaging Poly Bottle
Quantity 1 kg
Specific Gravity 1.137
Physical Form Liquid
Chemical Name or Material Triethyl Citrate FCC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.