Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl methanetricarboxylate, 98%, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
Packaging:
Glass bottle
6279-86-3
C10H16O6
232.23
AGZPNUZBDCYTBB-UHFFFAOYSA-N
tricarbethoxymethane,triethylmethanetricarboxylate,methanetricarboxylic acid, triethyl ester,2-ethoxycarbonyl-malonic acid diethyl ester,methanetricarboxylic acid triethyl ester,carboxymalonic acid triethyl ester,diethyl 2-ethoxycarbonyl malonate,diethyl 2-ethoxycarbonyl propane-1,3-dioate,pubchem3106,triethoxycarbonylmethane
80471
triethyl methanetricarboxylate
CCOC(=O)C(C(=O)OCC)C(=O)OCC
6279-86-3
C10H16O6
232.23
AGZPNUZBDCYTBB-UHFFFAOYSA-N
tricarbethoxymethane,triethylmethanetricarboxylate,methanetricarboxylic acid, triethyl ester,2-ethoxycarbonyl-malonic acid diethyl ester,methanetricarboxylic acid triethyl ester,carboxymalonic acid triethyl ester,diethyl 2-ethoxycarbonyl malonate,diethyl 2-ethoxycarbonyl propane-1,3-dioate,pubchem3106,triethoxycarbonylmethane
80471
triethyl methanetricarboxylate
CCOC(=O)C(C(=O)OCC)C(=O)OCC
Infrared Spectrum Authentic
Color Colorless to Yellow
Boiling Point 253.0°C
Packaging Glass bottle
Flash Point >110°C
Melting Point 29.0°C
Quantity 25mL
Refractive Index 1.4240 to 1.4260
Specific Gravity 1.1
Density 1.1000g/mL
Assay Percent Range 98%
Linear Formula CH(CO2C2H5)3
Beilstein 02,81
Solubility Information Solubility in water: immiscible
Formula Weight 232.23
Physical Form Liquid After Melting
Percent Purity 98%
Chemical Name or Material Triethyl methanetricarboxylate
Show More Show Less