Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethyl Orthoacetate 99%, Thermo Scientific™

Catalog No. AC421681000
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC421681000 100 g
AC421685000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC421681000 Supplier Thermo Scientific Chemicals Supplier No. 421681000

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-39-7
Molecular Formula C8H18O3
Molecular Weight (g/mol) 162.229
InChI Key NDQXKKFRNOPRDW-UHFFFAOYSA-N
Synonym triethyl orthoacetate, ethyl orthoacetate, ethane, 1,1,1-triethoxy, orthoacetic acid, triethyl ester, orthoacetic acid triethyl ester, triethoxyethane, triethoxy ethane, triethy orthoacetate, triethyl ortho-aceate, 1,1-triethoxyethane
PubChem CID 66221
IUPAC Name 1,1,1-triethoxyethane
SMILES CCOC(C)(OCC)OCC

Specifications

Color Colorless
Density 0.89
Infrared Spectrum Authentic
Refractive Index 1.3950 to 1.3980 (20°C, 589nm)
Linear Formula CH3C(OC2H5)3
Quantity 100 g
Beilstein 02, 129
Fieser 03,300; 04,527; 06,607; 10,417
Formula Weight 162.23
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material Triethyl orthoacetate, 99%
Show More Show Less
Hazard Category MOISTURE SENSITIVE
EINECSNumber 201-112-4
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.