Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylaluminium, 0.6M solution in heptane, AcroSeal™, Acros Organics

Click to view available options
Quantity:
100mL
800mL
Packaging:
AcroSeal™ Glass Bottle
97-93-8,142-82-5
C6H15Al
114.17
MFCD00009015
VOITXYVAKOUIBA-UHFFFAOYSA-N
triethylaluminum,triethylaluminium,aluminum, triethyl,triethylalane,unii-h426e9h3tt,triethylaluminum solution, 1.0 m in hexanes,triethylaluminum solution, 25 wt. % in toluene,triethyl-aluminum,triethyl aluminium,triethyl-aluminium
16682930
triethylalumane
CC[Al](CC)CC
97-93-8,142-82-5
C6H15Al
114.17
MFCD00009015
VOITXYVAKOUIBA-UHFFFAOYSA-N
triethylaluminum,triethylaluminium,aluminum, triethyl,triethylalane,unii-h426e9h3tt,triethylaluminum solution, 1.0 m in hexanes,triethylaluminum solution, 25 wt. % in toluene,triethyl-aluminum,triethyl aluminium,triethyl-aluminium
16682930
triethylalumane
CC[Al](CC)CC
Lewis acid
Packaging AcroSeal™ Glass Bottle
Density 0.6880g/mL
Color Colorless
Specific Gravity 0.688
Quantity 800mL
Linear Formula Al(CH2CH3)3
Merck Index 15, 326
Solubility Information Solubility in water: reacts. Other solubilities: miscible with saturated aliphatic and aromatic,hydrocarbons
Formula Weight 114.17
Physical Form Liquid
Chemical Name or Material Triethylaluminium, 0.6M Solution in Heptane
Show More Show Less

GHS H Statement
May cause drowsiness or dizziness. Causes severe skin burns and eye damage. Very toxic to aquatic life with long lasting effects. May be fatal if swallowed and enters airways. Catches fire spontaneously if exposed to air. Highly flammable liquid and vapor. Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting. Wear eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Keep away from heat/sparks/open flames/hot surfaces. - No smoking. Do not allow contact with air. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician. Do NOT induce vomiting. Avoid release to the environment. Store in a dry place. Store in a closed container.

GHS Signal Word: Danger

EINECSNumber : 202-619-3