Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylaluminum, 94%, Thermo Scientific Chemicals

Catalog No. aa4324322
Click to view available options
Quantity:
100 g

Catalyst for polymerization of olefins, intermediate

Triethylaluminum is used as a co-catalyst in the industrial production of polyethylene and for the production of medium chain alcohols. Used as a catalyst in Ziegler-Natta polymerization process for vinyl, olefin, diene polymerizations and linear oligomerization and cyclization of unsaturated hydrocarbons. It is also used as a catalyst to produce ethylene gas, chain growth of ethylene, longer chain aluminum alkyls, and in plating aluminum.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 97-93-8
Molecular Formula C6H15Al
Molecular Weight (g/mol) 114.168
MDL Number MFCD00009015
InChI Key VOITXYVAKOUIBA-UHFFFAOYSA-N
Synonym triethylaluminum, triethylaluminium, aluminum, triethyl, triethylalane, unii-h426e9h3tt, triethylaluminum solution, 1.0 m in hexanes, triethylaluminum solution, 25 wt. % in toluene, triethyl-aluminum, triethyl aluminum, triethyl-aluminum
PubChem CID 16682930
IUPAC Name triethylalumane
SMILES CC[Al](CC)CC

Specifications

Melting Point -50°C
Density 0.835 g/mL
Boiling Point 128°C to 130°C (50 mmHg)
Flash Point 18°C (−1°F)
Odor Characteristic
Packaging Cylinder
Quantity 100 g
UN Number UN3394
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 114.17
Percent Purity 94%
Physical Form Liquid
Chemical Name or Material Triethylaluminum
Show More Show Less
Hazard Category H225-H250-H260-H314-H335
Hazard Statement GHS H Statement
H225-H250-H260-H314
Highly flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
Precautionary Statement P210-P222-P223-P231+P232-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P302+P334-P304+P340-P305+P351+P338-P310-P335+P334-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, PYROPHORIC, WATER-REACTIVE
EINECSNumber 202-619-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.