Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylamine, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. p-7187922 Shop All MilliporeSigma Supelco Products
Click to view available options
Quantity:
1 mL
5 mL

Catalog No. 11-101-9672

Supplier: MilliporeSigma Supelco 734871ML

Only null left
Add to Cart
Add to Cart
Cleaning Products, Cosmetics, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 593°F
CAS 121-44-8
Vapor Pressure 51.75 mmHg (20°C)
Vapor Density 3.5 (vs. air)
Density 0.726 g/mL (at 25°C)
Flash Point -11°C
Physical Form Neat
Linear Formula (C2H5)3 N
MDL Number MFCD00009051
Melting Point -115°C
Boiling Point 88.8°C
Synonym N,N-Diethylethanamine
Type Analytical Standard
Quantity 1 mL
Formula Weight 101.19
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.