Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Triethylamine, 99.7%, extra pure

Catalog No. p-204002
Click to view available options
Quantity:
5 g
50 g
1 kg
2.5 kg
10 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 121-44-8 | C6H15N | 101.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-44-8
MDL Number MFCD00009051
InChI Key ZMANZCXQSJIPKH-UHFFFAOYSA-N
Synonym triethylamine, ethanamine, n,n-diethyl, diethylamino ethane, triaethylamin, triethylamin, trietilamina, triethyl amine, n,n,n-triethylamine, net3, diethylaminoethane
PubChem CID 8471
ChEBI CHEBI:35026
IUPAC Name N,N-diethylethanamine
SMILES CCN(CC)CC

Specifications

Melting Point -115.0°C
Density 0.7280g/mL
Boiling Point 90.0°C
Flash Point −11°C
Infrared Spectrum Authentic
Assay Percent Range 99.6% min. (GC)
Packaging Glass bottle
Molecular Formula C6H15N
Refractive Index 1.3990 to 1.4010
Linear Formula (C2H5)3N
Quantity 5 g
Beilstein 04, 99
Fieser 01,1198; 02,427; 04,527; 05,689; 07,385; 09,481; 11,553; 13,322; 15,336
Specific Gravity 0.728
Merck Index 15, 9831
Solubility Information Solubility in water: 133g/L (20°C). Other solubilities: miscible with ether, soluble in methanol, ethanol, 2-propanol, acetone, and methylenechloride
Molecular Weight (g/mol) 101.19
Viscosity 0.36 mPa.s (20°C)
Vapor Pressure 69mbar at 20°C
Formula Weight 101.19
Percent Purity 99.7%
Grade Extra Pure
Physical Form Liquid
Chemical Name or Material Triethylamine, 99.7%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapour.
Toxic if inhaled.
Toxic in contact with skin.
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Gently wash with plenty of soap and water.
IF IN EYES: Rinse cautiously wit
EINECSNumber 204-469-4
RTECSNumber YE0175000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.